Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ttc23
Entrez Gene ID
67009
Gene Name
tetratricopeptide repeat domain 23
Gene Aliases
1600012K10Rik
Chromosome Location
Chr.7: 67647410 - 67730060 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001168475.1 NP_001161947.1 6 - 7 1221 93
NM_001168476.1 NP_001161948.1 5 - 6 1021 93
GenBank mRNA AK005431.1 - 5 - 6 982 93
AK086049.1 - - 1288 93
AK141754.1 - 6 - 7 1207 93


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK011807.1 - Chr. - - [-] on Build 93

View Full Product Details