The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026810.2 | NP_081086.2 | 18 - 19 | 2194 | 103 |
GenBank mRNA | AF250844.1 | - | 18 - 19 | 2110 | 103 |
AK004105.1 | - | 9 - 10 | 1284 | 103 | |
AK145646.1 | - | 18 - 19 | 2141 | 103 | |
AK163861.1 | - | 17 - 18 | 3360 | 103 | |
AK168849.1 | - | 18 - 19 | 2133 | 103 | |
BC021815.1 | - | 18 - 19 | 2148 | 103 |