The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_026824.4 | NP_081100.2 | 13 - 14 | 1550 | 91 |
GenBank mRNA | AK004041.1 | - | 12 - 13 | 1073 | 91 |
AK037538.1 | - | 11 - 12 | 2687 | 91 | |
AK049333.1 | - | - | 421 | 91 | |
AK088243.1 | - | 13 - 14 | 1551 | 91 | |
AK147967.1 | - | 13 - 14 | 1750 | 91 | |
AK148307.1 | - | 13 - 14 | 1750 | 91 | |
BC019480.1 | - | 11 - 12 | 1034 | 91 |