The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159905.1 | NP_001153377.1 | 5 - 6 | 518 | 83 |
NM_001159906.1 | NP_001153378.1 | 5 - 6 | 515 | 83 | |
NM_026846.3 | NP_081122.2 | 4 - 5 | 552 | 83 | |
GenBank mRNA | AK004332.1 | - | 4 - 5 | 542 | 83 |
AK151430.1 | - | 4 - 5 | 554 | 83 | |
BC011495.1 | - | 4 - 5 | 577 | 83 | |
CX217579.1 | - | 5 - 6 | 521 | 83 |