The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027212.2 | NP_081488.1 | 2 - 3 | 398 | 75 |
GenBank mRNA | AK007739.1 | - | 2 - 3 | 391 | 75 |
AK011104.1 | - | 2 - 3 | 399 | 75 | |
AK146250.1 | - | 2 - 3 | 403 | 75 | |
BC020113.1 | - | 2 - 3 | 386 | 75 |