The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159349.1 | NP_001152821.1 | 5 - 6 | 678 | 93 |
NM_027226.4 | NP_081502.2 | 5 - 6 | 705 | 93 | |
GenBank mRNA | AK014447.1 | - | 5 - 6 | 676 | 93 |
AK029785.1 | - | 5 - 6 | 650 | 93 | |
AK050448.1 | - | 5 - 6 | 664 | 93 | |
AK050479.1 | - | 5 - 6 | 677 | 93 | |
AK077184.1 | - | 6 - 7 | 613 | 93 | |
AK139638.1 | - | 5 - 6 | 674 | 93 | |
AK157602.1 | - | 5 - 6 | 677 | 93 | |
AK161252.1 | - | 5 - 6 | 683 | 93 | |
AK165633.1 | - | 5 - 6 | 659 | 93 | |
AK166032.1 | - | 5 - 6 | 677 | 93 | |
AK170511.1 | - | 5 - 6 | 677 | 93 | |
AK211641.1 | - | 3 - 4 | 270 | 93 | |
BC028253.1 | - | 5 - 6 | 662 | 93 |