The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_027874.2 | NP_082150.1 | 8 - 9 | 1517 | 60 |
GenBank mRNA | AK004606.1 | - | 8 - 9 | 1277 | 60 |
AK146600.1 | - | 8 - 9 | 1507 | 60 | |
AK150387.1 | - | 8 - 9 | 1246 | 60 | |
AK150815.1 | - | 8 - 9 | 1244 | 60 | |
AK178798.1 | - | 1 - 2 | 55 | 60 | |
AK181467.1 | - | 1 - 2 | 55 | 60 | |
AK184083.1 | - | 1 - 2 | 55 | 60 | |
AK187539.1 | - | 1 - 2 | 55 | 60 | |
AK208057.1 | - | 1 - 2 | 55 | 60 | |
AK210596.1 | - | 1 - 2 | 55 | 60 |