The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_028390.3 | NP_082666.2 | 15 - 16 | 2811 | 137 |
GenBank mRNA | AK045703.1 | - | 12 - 13 | 1775 | 137 |
AK046102.1 | - | 4 - 5 | 597 | 137 | |
BC030489.1 | - | 8 - 9 | 1153 | 137 | |
BC032164.1 | - | 15 - 16 | 2829 | 137 |