The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252534.1 | NP_001239463.1 | 4 - 5 | 826 | 109 |
NM_001252536.1 | NP_001239465.1 | 5 - 6 | 550 | 109 | |
NM_001252537.1 | NP_001239466.1 | 4 - 5 | 826 | 109 | |
NM_001252538.1 | NP_001239467.1 | 4 - 5 | 516 | 109 | |
NM_028412.2 | NP_082688.1 | 4 - 5 | 468 | 109 | |
GenBank mRNA | AJ575057.1 | - | 2 - 3 | 137 | 109 |
AK089986.1 | - | 4 - 5 | 829 | 109 | |
AK137961.1 | - | 4 - 5 | 432 | 109 | |
AK159103.1 | - | 4 - 5 | 804 | 109 | |
AK165687.1 | - | 5 - 6 | 504 | 109 | |
BC018483.1 | - | 4 - 5 | 432 | 109 | |
BC072624.1 | - | - | 671 | 109 |