The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001252534.1 | NP_001239463.1 | 6 - 7 | 1148 | 64 |
NM_001252536.1 | NP_001239465.1 | 7 - 8 | 872 | 64 | |
NM_001252537.1 | NP_001239466.1 | 6 - 7 | 1133 | 64 | |
NM_001252538.1 | NP_001239467.1 | 6 - 7 | 823 | 64 | |
NM_028412.2 | NP_082688.1 | 6 - 7 | 790 | 64 | |
GenBank mRNA | AJ575057.1 | - | 4 - 5 | 444 | 64 |
AK089986.1 | - | 6 - 7 | 1136 | 64 | |
AK137961.1 | - | 6 - 7 | 739 | 64 | |
AK159103.1 | - | 6 - 7 | 1126 | 64 | |
AK165687.1 | - | 7 - 8 | 826 | 64 | |
BC018483.1 | - | 6 - 7 | 754 | 64 | |
BC072624.1 | - | - | 993 | 64 |