Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Ciz1
Entrez Gene ID
68379
Gene Name
CDKN1A interacting zinc finger protein 1
Gene Aliases
0610038H21Rik, 2900056O04Rik, LSFR1, ZNF356
Chromosome Location
Chr.2: 32363006 - 32378313 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001252534.1 NP_001239463.1 10 - 11 1781 87
NM_001252536.1 NP_001239465.1 11 - 12 1493 87
NM_001252537.1 NP_001239466.1 10 - 11 1754 87
NM_001252538.1 NP_001239467.1 10 - 11 1444 87
NM_028412.2 NP_082688.1 9 - 10 1558 87
GenBank mRNA AJ575057.1 - 8 - 9 1065 87
AK089986.1 - 10 - 11 1757 87
AK137961.1 - 10 - 11 1360 87
AK165687.1 - 11 - 12 1447 87
BC018483.1 - 9 - 10 1522 87
BC072624.1 - - 1626 87
CX217150.1 - 2 - 3 269 87


Non Gene Transcripts

Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA AK002799.1 - Chr. - - [-] on Build 87

View Full Product Details