The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001146023.1 | NP_001139495.1 | 6 - 7 | 764 | 71 |
GenBank mRNA | AK003496.1 | - | 5 - 6 | 688 | 71 |
AK080790.1 | - | 5 - 6 | 856 | 71 | |
AK171108.1 | - | 4 - 5 | 1130 | 71 | |
BC023421.1 | - | 3 - 4 | 382 | 71 | |
BC058765.1 | - | 5 - 6 | 821 | 71 |