The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_029788.5 | NP_084064.1 | 1 - 2 | 144 | 108 |
GenBank mRNA | AK002624.1 | - | 1 - 2 | 108 | |
AK036401.1 | - | 1 - 2 | 145 | 108 | |
AK153293.1 | - | 1 - 2 | 113 | 108 | |
BC050796.1 | - | 1 - 2 | 112 | 108 |