Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Rpe65
Entrez Gene ID
19892
Gene Name
retinal pigment epithelium 65
Gene Aliases
65kDa, A930029L06Rik, LCA2, Mord1, RP20, rd12
Chromosome Location
Chr.3: 159599075 - 159625307 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_029987.2 NP_084263.2 4 - 5 396 95
GenBank mRNA AF410461.1 - 3 - 4 344 95
AK020906.1 - 4 - 5 395 95
BC090656.1 - 4 - 5 372 95
BC130028.1 - 3 - 4 348 95
CJ133618.1 - 4 - 5 397 95

View Full Product Details