The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001277891.1 | NP_001264820.1 | 1 - 2 | 114 | 71 |
NM_001277893.1 | NP_001264822.1 | 1 - 2 | 114 | 71 | |
NM_033571.3 | NP_291049.1 | 1 - 2 | 114 | 71 | |
GenBank mRNA | AF367709.1 | - | 1 - 2 | 95 | 71 |
AF367710.1 | - | 1 - 2 | 61 | 71 | |
AF367711.1 | - | 1 - 2 | 61 | 71 | |
BC139092.1 | - | 1 - 2 | 114 | 71 | |
BC139097.1 | - | 1 - 2 | 114 | 71 |