Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ankrd6
Entrez Gene ID
140577
Gene Name
ankyrin repeat domain 6
Gene Aliases
-
Chromosome Location
Chr.4: 32804027 - 32950884 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001012450.1 NP_001012453.1 3 - 4 634 65
NM_001012451.1 NP_001012454.1 2 - 3 345 65
NM_080471.3 NP_536719.2 2 - 3 474 65
GenBank mRNA AK133551.1 - 1 - 2 361 65
AK135024.1 - 2 - 3 329 65
AK173070.1 - 2 - 3 385 65
AY026320.2 - 1 - 2 266 65
BC065177.1 - 3 - 4 584 65
CF531081.1 - 2 - 3 474 65
CF533090.1 - 2 - 3 360 65

View Full Product Details