Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Ppap2b
Entrez Gene ID
67916
Gene Name
phosphatidic acid phosphatase type 2B
Gene Aliases
1110003O22Rik, 2610002D05Rik, AV025606, D4Bwg0538e, D4Bwg1535e, Lpp3, Ppab2b
Chromosome Location
Chr.4: 105157347 - 105232767 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_080555.2 NP_542122.1 1 - 2 621 66
GenBank mRNA AK011276.1 - 1 - 2 620 66
AK076183.1 - 1 - 2 317 66
AK159136.1 - 1 - 2 707 66
AK160056.1 - 1 - 2 521 66
BC005558.1 - 1 - 2 608 66

View Full Product Details