The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130163.1 | NP_001123635.1 | 10 - 11 | 1950 | 79 |
NM_001130164.1 | NP_001123636.1 | 2 - 3 | 303 | 79 | |
NM_001130165.1 | NP_001123637.1 | 10 - 11 | 2045 | 79 | |
NM_001130166.1 | NP_001123638.1 | 13 - 14 | 2390 | 79 | |
NM_130885.2 | NP_570955.1 | 11 - 12 | 2031 | 79 | |
GenBank mRNA | AF324899.1 | - | 11 - 12 | 2030 | 79 |
AF333985.1 | - | 10 - 11 | 1950 | 79 | |
AK158703.1 | - | 13 - 14 | 2372 | 79 | |
AW106090.1 | - | 2 - 3 | 316 | 79 | |
BC003927.1 | - | 4 - 5 | 540 | 79 | |
BC089183.1 | - | 10 - 11 | 2040 | 79 | |
BC098491.1 | - | 12 - 13 | 2292 | 79 |