The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001130163.1 | NP_001123635.1 | 11 - 12 | 2097 | 95 |
NM_001130164.1 | NP_001123636.1 | 3 - 4 | 450 | 95 | |
NM_001130165.1 | NP_001123637.1 | 11 - 12 | 2192 | 95 | |
NM_001130166.1 | NP_001123638.1 | 14 - 15 | 2537 | 95 | |
NM_130885.2 | NP_570955.1 | 12 - 13 | 2178 | 95 | |
GenBank mRNA | AF324899.1 | - | 12 - 13 | 2177 | 95 |
AF333985.1 | - | 11 - 12 | 2097 | 95 | |
AK158703.1 | - | 14 - 15 | 2519 | 95 | |
BC003927.1 | - | 5 - 6 | 687 | 95 | |
BC089183.1 | - | 11 - 12 | 2187 | 95 | |
BC098491.1 | - | 13 - 14 | 2439 | 95 |