The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_177409.3 | NP_803128.1 | 1 - 2 | 228 | 100 |
GenBank mRNA | AK082751.1 | - | 1 - 2 | 229 | 100 |
AY029530.1 | - | 1 - 2 | 165 | 100 | |
BC018212.1 | - | 1 - 2 | 213 | 100 |