The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133355.1 | NP_579933.1 | 1 - 2 | 71 | 59 |
GenBank mRNA | AF099933.2 | - | 1 - 2 | 71 | 59 |