Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Grid2ip
Entrez Gene ID
170935
Gene Name
glutamate receptor, ionotropic, delta 2 (Grid2) interacting protein 1
Gene Aliases
-
Chromosome Location
Chr.5: 143357338 - 143391798 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159321.1 NP_001152793.1 6 - 7 1259 96
NM_133355.1 NP_579933.1 5 - 6 563 96
GenBank mRNA AF099933.2 - 5 - 6 563 96
BC075624.1 - - 615 96
DQ193535.1 - 6 - 7 1259 96

View Full Product Details