Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Lad1
Entrez Gene ID
16763
Gene Name
ladinin
Gene Aliases
-
Chromosome Location
Chr.1: 135818598 - 135833341 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133664.3 NP_598425.2 8 - 9 1888 71
GenBank mRNA AK083526.1 - 8 - 9 1889 71
BC016257.1 - 8 - 9 1559 71
BC025580.1 - 8 - 9 1792 71
BC031131.1 - 8 - 9 1735 71

View Full Product Details