The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133666.3 | NP_598427.1 | 9 - 10 | 1442 | 73 |
GenBank mRNA | AA223049.1 | - | 2 - 3 | 180 | 73 |
AK005021.1 | - | 9 - 10 | 1390 | 73 | |
AK075692.1 | - | 9 - 10 | 1387 | 73 | |
AK151455.1 | - | 9 - 10 | 1353 | 73 | |
AW113957.1 | - | 1 - 2 | 73 | ||
BC014818.1 | - | 9 - 10 | 1412 | 73 | |
BC041682.1 | - | 9 - 10 | 1345 | 73 |