The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001159632.1 | NP_001153104.1 | 1 - 2 | 98 | 80 |
NM_133699.2 | NP_598460.1 | 1 - 2 | 98 | 80 | |
GenBank mRNA | AB088357.1 | - | 1 - 2 | 83 | 80 |
BC056636.1 | - | 1 - 2 | 71 | 80 | |
BC064071.1 | - | 1 - 2 | 33 | 80 |