The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133704.4 | NP_598465.2 | 4 - 5 | 650 | 68 |
GenBank mRNA | AK007341.1 | - | 4 - 5 | 590 | 68 |
AK044615.1 | - | 4 - 5 | 585 | 68 | |
AK082516.1 | - | 4 - 5 | 650 | 68 | |
BC010189.1 | - | 4 - 5 | 379 | 68 | |
BC138969.1 | - | 3 - 4 | 564 | 68 | |
BC138971.1 | - | 3 - 4 | 564 | 68 | |
BC145478.1 | - | 3 - 4 | 566 | 68 |