Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Gmppa
Entrez Gene ID
69080
Gene Name
GDP-mannose pyrophosphorylase A
Gene Aliases
1810012N01Rik
Chromosome Location
Chr.1: 75435912 - 75443176 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133708.3 NP_598469.1 8 - 9 846 72
GenBank mRNA AK007463.1 - 8 - 9 833 72
AK144311.1 - 8 - 9 1111 72
AK150473.1 - 9 - 10 982 72
BC008116.1 - 8 - 9 805 72

View Full Product Details