The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133727.2 | NP_598488.2 | 3 - 4 | 472 | 112 |
GenBank mRNA | AK009746.1 | - | 3 - 4 | 453 | 112 |
BC018316.1 | - | 3 - 4 | 440 | 112 |