Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- Dcaf11
- Entrez Gene ID
-
28199
- Gene Name
- DDB1 and CUL4 associated factor 11
- Gene Aliases
-
0710008A13Rik, C76035, D14Ucla1, Dacf11, GLO14, Wdr23
- Chromosome Location
-
Chr.14: 55559937 - 55570065 on Build GRCm38
Non Gene Transcripts
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
IMAGE Clone ID |
Transcript Location |
Amplicon Length |
GenBankmRNA |
AK003028.1 |
- |
|
|
|
Chr. - - [-] on Build |
109 |
View Full Product Details