Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Gpn1
Entrez Gene ID
74254
Gene Name
GPN-loop GTPase 1
Gene Aliases
2410004J02Rik, AI449615, MBDIN, NTPBP, Xab1
Chromosome Location
Chr.5: 31494752 - 31511631 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133756.4 NP_598517.1 12 - 13 945 101
GenBank mRNA AK010393.1 - 12 - 13 958 101
AK077645.1 - 12 - 13 946 101
AK145957.1 - - 942 101
AK146686.1 - - 957 101
BC020174.1 - 12 - 13 945 101
BC096466.1 - 12 - 13 938 101

View Full Product Details