The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133764.3 | NP_598525.1 | 1 - 2 | 283 | 63 |
GenBank mRNA | AK002260.1 | - | 1 - 2 | 151 | 63 |
AK077313.1 | - | 1 - 2 | 155 | 63 | |
BC010790.1 | - | 1 - 2 | 237 | 63 | |
BI134693.1 | - | 3 - 4 | 63 | ||
BY296901.1 | - | 1 - 2 | 283 | 63 |