The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133801.2 | NP_598562.1 | 3 - 4 | 296 | 133 |
GenBank mRNA | AK013002.1 | - | 3 - 4 | 310 | 133 |
AK078048.1 | - | 3 - 4 | 309 | 133 | |
AK168236.1 | - | 3 - 4 | 309 | 133 | |
AK168243.1 | - | 3 - 4 | 309 | 133 | |
BC003336.1 | - | 3 - 4 | 294 | 133 | |
BC023081.1 | - | 3 - 4 | 296 | 133 | |
BC031123.1 | - | 3 - 4 | 426 | 133 |