Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Clp1
Entrez Gene ID
98985
Gene Name
CLP1, cleavage and polyadenylation factor I subunit
Gene Aliases
AI462438, Heab
Chromosome Location
Chr.2: 84723122 - 84727268 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_133840.2 NP_598601.1 1 - 2 106 94
GenBank mRNA AK077647.1 - 1 - 2 83 94
AK145209.1 - - 81 94
AK169709.1 - 1 - 2 115 94
BC003237.1 - 1 - 2 106 94

View Full Product Details