Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Magi3
Entrez Gene ID
99470
Gene Name
membrane associated guanylate kinase, WW and PDZ domain containing 3
Gene Aliases
4732496O19Rik, 6530407C02Rik, AA407180, AI120132, mKIAA1634
Chromosome Location
Chr.3: 104013259 - 104220406 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001159354.1 NP_001152826.1 15 - 16 3073 85
NM_133853.3 NP_598614.1 15 - 16 3073 85
GenBank mRNA AF213258.1 - 15 - 16 2694 85
AK030083.1 - 15 - 16 2477 85
AK173225.1 - 10 - 11 1625 85

View Full Product Details