The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133952.2 | NP_598713.2 | 4 - 5 | 527 | 102 |
GenBank mRNA | AK028912.1 | - | 4 - 5 | 513 | 102 |
AK047292.1 | - | 4 - 5 | 474 | 102 | |
AK047476.1 | - | 4 - 5 | 474 | 102 | |
AK088034.1 | - | 4 - 5 | 479 | 102 | |
AK166809.1 | - | 4 - 5 | 510 | 102 | |
BC004717.1 | - | 4 - 5 | 516 | 102 |