The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_133964.2 | NP_598725.2 | 1 - 2 | 151 | 65 |
GenBank mRNA | AK004052.1 | - | 1 - 2 | 117 | 65 |
AK154104.1 | - | 1 - 2 | 152 | 65 | |
BC002295.1 | - | 1 - 2 | 68 | 65 |