The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001142580.1 | NP_001136052.1 | 8 - 9 | 730 | 64 |
NM_001142581.1 | NP_001136053.1 | 7 - 8 | 673 | 64 | |
NM_134044.3 | NP_598805.2 | 7 - 8 | 687 | 64 | |
GenBank mRNA | AK038165.1 | - | 8 - 9 | 698 | 64 |
AK046322.1 | - | 8 - 9 | 701 | 64 | |
AK049643.1 | - | 8 - 9 | 677 | 64 | |
AK079055.1 | - | 8 - 9 | 677 | 64 | |
AK082261.1 | - | 8 - 9 | 658 | 64 | |
AK145043.1 | - | - | 713 | 64 | |
BC016646.1 | - | 7 - 8 | 596 | 64 | |
BC023716.1 | - | 7 - 8 | 698 | 64 |