The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001166456.1 | NP_001159928.1 | 2 - 3 | 428 | 93 |
NM_001166458.1 | NP_001159930.1 | 2 - 3 | 635 | 93 | |
NM_134086.4 | NP_598847.2 | 2 - 3 | 1121 | 93 | |
GenBank mRNA | AF184240.2 | - | 1 - 2 | 213 | 93 |
AK029189.1 | - | 2 - 3 | 298 | 93 | |
AK034130.1 | - | 2 - 3 | 1123 | 93 | |
AK035098.1 | - | 1 - 2 | 140 | 93 | |
AK047459.1 | - | 2 - 3 | 652 | 93 | |
AK049173.1 | - | 2 - 3 | 335 | 93 | |
AK049294.1 | - | 2 - 3 | 401 | 93 | |
AK050914.1 | - | 2 - 3 | 336 | 93 | |
AK081724.1 | - | 2 - 3 | 637 | 93 | |
AK082683.1 | - | 2 - 3 | 336 | 93 | |
AK162612.1 | - | 2 - 3 | 486 | 93 | |
BC030378.1 | - | 2 - 3 | 337 | 93 | |
BC066815.1 | - | 2 - 3 | 298 | 93 |