The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134090.2 | NP_598851.2 | 1 - 2 | 236 | 104 |
GenBank mRNA | AK136991.1 | - | 1 - 2 | 220 | 104 |
BC011472.1 | - | 1 - 2 | 176 | 104 | |
BC024420.1 | - | 1 - 2 | 169 | 104 |