The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_134122.2 | NP_598883.1 | 3 - 4 | 572 | 63 |
GenBank mRNA | BC021750.1 | - | 3 - 4 | 575 | 63 |
BC026576.1 | - | 3 - 4 | 544 | 63 |