Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Vwa7
Entrez Gene ID
27762
Gene Name
von Willebrand factor A domain containing 7
Gene Aliases
C6orf27, D17H6S56E-3, G7c, NG37
Chromosome Location
Chr.17: 35016357 - 35026741 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138582.1 NP_613048.1 10 - 11 1710 108
GenBank mRNA AF134319.1 - 10 - 11 1710 108
BC137792.1 - 10 - 11 1651 108
BC144911.1 - 10 - 11 1588 108

View Full Product Details