Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Bspry
Entrez Gene ID
192120
Gene Name
B-box and SPRY domain containing
Gene Aliases
-
Chromosome Location
Chr.4: 62479995 - 62497300 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138653.1 NP_619594.1 2 - 3 467 83
GenBank mRNA AJ276690.1 - 2 - 3 467 83
AK035588.1 - 2 - 3 473 83
AK160930.1 - 2 - 3 473 83
BC139079.1 - 2 - 3 483 83
BC139080.1 - 2 - 3 483 83
BC145535.1 - 2 - 3 483 83

View Full Product Details