Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Socs7
Entrez Gene ID
192157
Gene Name
suppressor of cytokine signaling 7
Gene Aliases
2310063P06Rik, C85125, Nap4
Chromosome Location
Chr.11: 97362296 - 97398542 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_138657.3 NP_619598.1 6 - 7 1442 85
GenBank mRNA AF424814.1 - 6 - 7 1443 85
AK147580.1 - 6 - 7 1540 85
AK147650.1 - 6 - 7 1540 85

View Full Product Details