The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_138955.3 | NP_620405.3 | 14 - 15 | 1962 | 83 |
GenBank mRNA | AF378330.1 | - | 13 - 14 | 1793 | 83 |
AF411084.1 | - | 14 - 15 | 1840 | 83 | |
AF425077.1 | - | 14 - 15 | 1842 | 83 | |
AJ426047.2 | - | 14 - 15 | 1841 | 83 | |
AK043147.1 | - | 14 - 15 | 1963 | 83 | |
AK157317.1 | - | 14 - 15 | 2287 | 83 | |
AY040865.1 | - | 14 - 15 | 1806 | 83 | |
BC016200.1 | - | 14 - 15 | 1842 | 83 | |
BC023077.1 | - | 8 - 9 | 1050 | 83 | |
BC059033.1 | - | 5 - 6 | 1135 | 83 |