Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Cdkal1
Entrez Gene ID
68916
Gene Name
CDK5 regulatory subunit associated protein 1-like 1
Gene Aliases
1190005B03Rik, 6620401C13Rik
Chromosome Location
Chr.13: 29191751 - 29855684 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AK018346.1 - 4 - 5 411 63
AK041864.1 - 4 - 5 407 63
AK082629.1 - 4 - 5 414 63
AK158546.1 - 4 - 5 404 63
AK169761.1 - 4 - 5 411 63
BC016073.1 - 4 - 5 355 63

View Full Product Details