Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
Slc9a8
Entrez Gene ID
77031
Gene Name
solute carrier family 9 (sodium/hydrogen exchanger), member 8
Gene Aliases
1200006P13Rik, 6430709P13Rik, AI182282, NHE-8, NHE8
Chromosome Location
Chr.2: 167420056 - 167477000 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
GenBank mRNA AB089793.1 - 12 - 13 1183 100
AF482993.1 - 11 - 12 1147 100
AK049410.1 - 12 - 13 1179 100
AK051791.1 - 11 - 12 1086 100
AK129013.1 - 3 - 4 400 100
AK143281.1 - 12 - 13 1152 100
AK173069.1 - 12 - 13 1519 100
BC030879.1 - 12 - 13 1182 100
BC034508.1 - 5 - 6 583 100
BC058947.1 - 12 - 13 1153 100

View Full Product Details