The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_009641.1 | NP_033771.1 | 8 - 9 | 1364 | 60 |
GenBank mRNA | AF113707.1 | - | 8 - 9 | 1364 | 60 |
AK132988.1 | - | - | 1489 | 60 | |
AK154379.1 | - | - | 1489 | 60 | |
BC129965.1 | - | 8 - 9 | 1364 | 60 | |
BC129966.1 | - | 8 - 9 | 1364 | 60 |