Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
Anxa10
Entrez Gene ID
26359
Gene Name
annexin A10
Gene Aliases
-
Chromosome Location
Chr.8: 62057042 - 62123193 on Build GRCm38
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136089.2 NP_001129561.1 7 - 8 701 71
NM_011922.3 NP_036052.2 6 - 7 641 71
GenBank mRNA AJ238978.1 - 7 - 8 601 71
AK008772.1 - 7 - 8 624 71
AK008930.1 - 7 - 8 704 71
AK018597.1 - 7 - 8 658 71
AK020288.1 - 6 - 7 566 71
AK131880.1 - 7 - 8 621 71
AK168715.1 - 7 - 8 605 71
BC125319.1 - 7 - 8 573 71
BC132207.1 - 7 - 8 573 71

View Full Product Details