The assay probe spans an exon junction. This assay will detect gDNA.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038587.4 | NP_001033676.2 | 8 - 9 | 2620 | 61 |
NM_001146296.1 | NP_001139768.1 | 8 - 9 | 2612 | 61 | |
NM_019655.3 | NP_062629.3 | 8 - 9 | 2534 | 61 | |
GenBank mRNA | AF052506.1 | - | 8 - 9 | 2447 | 61 |
AF291050.1 | - | 8 - 9 | 2441 | 61 | |
AF291875.1 | - | 7 - 8 | 1009 | 61 | |
AF291876.1 | - | 8 - 9 | 2519 | 61 | |
AF291877.1 | - | 7 - 8 | 1087 | 61 | |
AK076413.1 | - | 8 - 9 | 2570 | 61 | |
AK089451.1 | - | 8 - 9 | 2583 | 61 | |
AK147614.1 | - | 8 - 9 | 2583 | 61 | |
AK156093.1 | - | 7 - 8 | 1101 | 61 | |
AK172523.1 | - | 7 - 8 | 1101 | 61 | |
BC042505.1 | - | 8 - 9 | 2468 | 61 | |
CT010323.1 | - | 8 - 9 | 2441 | 61 |