The assay probe spans an exon junction.
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | Amplicon Length | |
---|---|---|---|---|---|
RefSeq | NM_001038587.4 | NP_001033676.2 | 11 - 12 | 2966 | 58 |
NM_001146296.1 | NP_001139768.1 | 11 - 12 | 2958 | 58 | |
NM_019655.3 | NP_062629.3 | 11 - 12 | 2880 | 58 | |
GenBank mRNA | AF052506.1 | - | 11 - 12 | 2793 | 58 |
AF291050.1 | - | 11 - 12 | 2787 | 58 | |
AF291875.1 | - | 10 - 11 | 1355 | 58 | |
AF291876.1 | - | 11 - 12 | 2865 | 58 | |
AF291877.1 | - | 10 - 11 | 1433 | 58 | |
AK076413.1 | - | 11 - 12 | 2917 | 58 | |
AK089451.1 | - | 11 - 12 | 2929 | 58 | |
AK147614.1 | - | 11 - 12 | 2929 | 58 | |
AK156093.1 | - | 10 - 11 | 1447 | 58 | |
AK172523.1 | - | 10 - 11 | 1447 | 58 | |
BC042505.1 | - | 11 - 12 | 2814 | 58 | |
CT010323.1 | - | 11 - 12 | 2787 | 58 |